1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose ≥92.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Diacetone-D-galactose, 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, ((3aR,5R,5aS,8aS,8bR)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-5-yl)methanol
CAS Number:4064-06-6
:
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00063225
Boiling Pt:133°C
Molecular Weight:260.29 g/mol
EINECS:223-771-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 92.0 %
- Specific rotation [a]20/D:-48.0 to -52.0 deg (C=5, MeOH)
Specification Test Results
Related Information
Frequently Bought Together