Phenyl-1,4-dihydroxy-2-naphthoate ≥98.0% (by HPLC)
Catalog # TCIAD2297-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:153.0 to 157.0 deg-C
Specification Test Results
About this item
:
:280.28 g/mol
EINECS:259-419-4
CAS Number:54978-55-1
Formula:C₁₇H₁₂O₄
MDL Number:MFCD00075770