Diethylene glycol bis(2-chloroethyl)ether ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Diethylene Glycol Bis(2-chloroethyl) Ether, Bis[2-(2-chloroethoxy)ethyl] ether
CAS Number:638-56-2
:
Formula:C₈H₁₆Cl₂O₃
MDL Number:MFCD00054473
:231.11 g/mol
UN:2810
ADR:6.1,III
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together