Order Entry
Denmark
ContactUsLinkComponent
Sodium dodecylbenzenesulphonate (mixture) ≥95.0% (by titrimetric analysis), soft type
Irritant
Sodium dodecylbenzenesulphonate (mixture) ≥95.0% (by titrimetric analysis), soft type
Supplier:  TCI
Certificates
Irritant
Sodium dodecylbenzenesulphonate (mixture) ≥95.0% (by titrimetric analysis), soft type
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Phase splitting method):
    min. 95.0 %
  • Solubility in Water:
    almost transparency
  • Petroleum ether soluble matter:
    max. 2.0 %

Specification Test Results

Related Information

Frequently Bought Together