

3,5-Dimethylheptane ≥99.0% (by GC)
TCIAD1208-0.5ML
: TCI
:
- Pk:0,5 mL
- Seq:2040
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 % (total of isomer)
:
:128.26 g/mol
:136°C
:23°C
:1920
:3,III
:GHS02
:GHS08
:926-82-9
:C₉H₂₀
:MFCD00048659