
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1201-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:189.0 to 193.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:310.36 g/mol
EINECS:213-878-7
:GHS07
CAS Number:1046-56-6
Formula:C₂₀H₁₄N₄
MDL Number:MFCD00006462