4,4'-Diantipyrylmethane monohydrate ≥98.0% (by titrimetric analysis) for Ti analysis
Catalog # TCIAD1029-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.000 area%
- Purity (Nonaqueous Titration):98.0 to 102.0 %
- Solubility in dilute HCl:almost transparency
- Suitability for Titanium-analysis:Abs min.0.28 (near 390nm) in the presence of Ti (1 ppm)
Specification Test Results
About this item
:
:388.47 g/mol (anhy)
EINECS:215-009-7
CAS Number:1251-85-0
Formula:C₂₃H₂₄N₄O₂·H₂O
MDL Number:MFCD00149122