Order Entry
Denmark
ContactUsLinkComponent
4-Diazodiphenylamine sulphate ≥98.0% (by titrimetric analysis)
4-Diazodiphenylamine sulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD0141-25G
Supplier:  TCI
CAS Number:  
4-Diazodiphenylamine sulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD0141-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :293.30 g/mol
EINECS:224-756-8
CAS Number:4477-28-5
Formula:C₁₂H₉N₃·H₂SO₄
MDL Number:MFCD00011895