2-Deoxy-D-galactose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1949-89-9
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00014649
Molecular Weight:164.16 g/mol
EINECS:217-765-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Melting point:106.0 to 114.0 deg-C
- Specific rotation [a]20/D:+55.0 to +62.0 deg (C=2,H2O)
Specification Test Results
Frequently Bought Together