N-Cbz-2-piperidinecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:28697-07-6
:
Formula:C₁₄H₁₇NO₄
MDL Number:MFCD01863568
:263.2900 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:78.0 to 82.0 deg-C
Specification Test Results
Frequently Bought Together