Z-trans-4-Hydroxy-L-prolinol ≥95.0% (by GC)
TCIAC3255-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Specific rotation [a]20/D:-42.0 to -46.0 deg (C=1, CHCL3)
:
:251.282 g/mol
:95687-41-5
:C₁₃H₁₇NO₄
:MFCD03427085