Order Entry
Denmark
ContactUsLinkComponent
6-Chlorochromone-2-carboxylic acid ≥98.0% (by GC, titration analysis)
6-Chlorochromone-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAC3174-1G
Supplier:  TCI
CAS Number:  
undefined
6-Chlorochromone-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAC3174-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
Melting Pt:269°C
 :GHS07
CAS Number:5006-45-1
Formula:C₁₀H₅ClO₄
MDL Number:MFCD00847039