1-(4-Chlorophenyl)cyclopentanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:80789-69-1
:
Formula:C₁₂H₁₃ClO₂
MDL Number:MFCD00001373
:224.6834 g/mol
EINECS:279-552-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
Frequently Bought Together