Order Entry
Denmark
ContactUsLinkComponent
 
 
5-Carboxyfluorescein, hydrate ≥97.0% (by HPLC, titration analysis)
  TCIAC2477-100MG
 :  TCI
 :  
 
5-Carboxyfluorescein, hydrate ≥97.0% (by HPLC, titration analysis)
  TCIAC2477-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 % (calcd.on anh.substance)
  • Water:
    max. 10.0 %

 

 

 :
 :376.32 g/mol (anhy)
 :GHS07
 :76823-03-5
 :C₂₁H₁₂O₇.xH₂O
 :MFCD00036874