Citalopram hydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:59729-32-7
:
Formula:C₂₀H₂₁FN₂O
MDL Number:MFCD02101306
Merck Index:14,02318
:405.31 g/mol
EINECS:261-890-6
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together