
Curcumin ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:458-37-7
:
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365
:368.39 g/mol
Merck Index:14,02673
EINECS:207-280-5
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:182.0 to 188.0 deg-C
Specification Test Results
Frequently Bought Together