
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
: TCI
:86604-75-3
:
:C₉H₁₂ClNO
:MFCD00277472
:222.11 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together