

1-Chloro-2-methylpentane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14753-05-0
:
Formula:C₆H₁₃Cl
MDL Number:MFCD00053236
:120.62 g/mol
Boiling Pt:120°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together