Order Entry
Denmark
ContactUsLinkComponent
 
Nɑ-Carbobenzoxy-DL-asparagine ≥99.0% (by titrimetric analysis)
 :  TCI
 
Nɑ-Carbobenzoxy-DL-asparagine ≥99.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together