N-Carbobenzoxy-DL-leucine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:3588-60-1
:
Formula:C₁₄H₁₉NO₄
MDL Number:MFCD00065129
:265.31 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:54.0 to 57.0 deg-C
- Specific rotation [a]20/D:-5.0 to 5.0 deg (C=2, ethanol)
Specification Test Results
Frequently Bought Together