N-Carbobenzoxy-DL-serine ≥98.0% (by titrimetric analysis)
Catalog # TCIAC0637-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:125.0 to 127.0 deg-C
Specification Test Results
About this item
:
:239.23 g/mol
EINECS:220-455-0
CAS Number:2768-56-1
Formula:C₁₁H₁₃NO₅
MDL Number:MFCD00063143