
tert-Butyl Hydrogen tetradecanedioate ≥95.0% (by GC, titration analysis)
Catalog # TCIAB5927-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g/mol
- Seq:9220
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:49.0 to 53.0 deg-C
Specification Test Results
About this item
:
:314.47 g/mol
:GHS07
CAS Number:234082-00-9
Formula:C₁₈H₃₄O₄
MDL Number:MFCD23136040