2,6-Bis[(2-hydroxyethyl)amino]toluene ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:149330-25-6
:
Formula:C₁₁H₁₈N₂O₂
MDL Number:MFCD08703311
:210.28 g/mol
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
Frequently Bought Together