(S)-alpha,alpha-Bis[3,5-bis(trifluoromethyl)phenyl]-2-pyrrolidinemethanol ≥98.0% (by GC, titration analysis)
Catalog # TCIAB5701-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
- Specific rotation [a]20/D:-50.0 to -56.0 deg (C=1, CHCL3)
Specification Test Results
About this item
:
:525.34 g/mol
CAS Number:848821-76-1
Formula:C₂₁H₁₅F₁₂NO
MDL Number:MFCD10566964