Order Entry
Denmark
ContactUsLinkComponent
 
Boc-6-Ahx-OSu ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
Boc-6-Ahx-OSu ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    88.0 to 92.0 deg-C

 

Frequently Bought Together