Biotin-PEG11-Amine ≥93.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1418022-42-0
:
Formula:C₃₄H₆₆N₄O₁₃S
:770.98 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 93.0 area%
- Elemental analysis (Nitrogen):6.50 to 8.00 %
Specification Test Results
Frequently Bought Together