4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1044795-04-1
:
Formula:C₃₄H₅₄O₂S₂
:558.92 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together