2-Butoxyethyl acetate 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Butylglycol acetate
- CAS Number:112-07-2
- Formula:C₈H₁₆O₃
- MDL Number:MFCD00009457
- Molecular Weight:160.21 g/mol
- Boiling Pt:193.5 °C (760 mmHg)
- Melting Pt:–64 °C
- Density:0.9385 g/cm³
- Flash Pt:76 °C
- Storage Temperature:Ambient
- EINECS:203-933-3
3 Options Available Below
Product Details & Documents
Synonyms: Butylglycol acetate
- CAS Number:112-07-2
- Formula:C₈H₁₆O₃
- MDL Number:MFCD00009457
- Molecular Weight:160.21 g/mol
- Boiling Pt:193.5 °C (760 mmHg)
- Melting Pt:–64 °C
- Density:0.9385 g/cm³
- Flash Pt:76 °C
- Storage Temperature:Ambient
- EINECS:203-933-3
Documents
ACRO40341
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4150 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤15 APHA
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



