Methyl undecanoate
Glass bottle, 10 mL
Catalog # ROTH4866.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Undecanoic acid methyl ester, Undecylic acid methyl ester
- CAS Number:1731-86-8
- Formula:C₁₂H₂₄O₂
- MDL Number:MFCD00008957
- Molecular Weight:200.32 g/mol
- EINECS:217-053-2
Specifications
Specifications
- Pack type:Glass bottle
- Pk:10 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



