Order Entry
Denmark
ContactUsLinkComponent
 
Bethanechol chloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
Bethanechol chloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    214.0 to 218.0 deg-C

 

Frequently Bought Together