Order Entry
Denmark
ContactUsLinkComponent
2-Bromo-3-(2-ethylhexyl)thiophene ≥98.0% (by GC)
2-Bromo-3-(2-ethylhexyl)thiophene ≥98.0% (by GC)
Supplier:  TCI
Certificates
2-Bromo-3-(2-ethylhexyl)thiophene ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together