
5'-Bromo-2'-hydroxy-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)
: TCI
:70978-54-0
:
:C₈H₆BrNO₄
:MFCD01631134
:260.05 g/mol
:GHS07
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
Frequently Bought Together