Bis[rhodium(alpha,alpha,alpha',alpha'-tetramethyl-1,3-benzenedipropionic acid)] ≥96.0% (by HPLC)
Catalog # TCIAB4549-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 96.0 area%
Specification Test Results
About this item
:
:758.48 g/mol
CAS Number:819050-89-0
Formula:C₃₂H₄₀O₈Rh₂
MDL Number:MFCD08457636