4-Bromopicolinic acid ≥98.0% (by GC, titration analysis)
: TCI
: 4-Bromopyridine-2-carboxylic acid
:30766-03-1
:
:C₆H₄BrNO₂
:MFCD03426665
:202.01 g/mol
:179°C
:GHS07
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together