(+)-Biotin-4-nitrophenyl ester ≥98.0% (by HPLC, total nitrogen)
TCIAB4009-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Melting point:163.0 to 167.0 deg-C
- Specific rotation [a]20/D:+48.0 to +52.0 deg (C=1, DMF, calcd.on anh.substance)
:
:365.40 g/mol
:33755-53-2
:C₁₆H₁₉N₃O₅S
:MFCD15141972