Order Entry
Denmark
ContactUsLinkComponent
Ethyl palmitate
Ethyl palmitate
Catalog # CAYM10008202-5
Supplier:  Cayman Chemical
CAS Number:  
Ethyl palmitate
Catalog # CAYM10008202-5
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :284.48 g/mol
Boiling Pt:342 °C (1013 hPa)
Melting Pt:24 °C
Density:0.86 g/cm³ (20 °C)
EINECS:211-064-6
CAS Number:628-97-7
Formula:C₁₈H₃₆O₂
MDL Number:MFCD00008996