2-Bromobenzophenone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13047-06-8
:
Formula:C₁₃H₉BrO
MDL Number:MFCD00032462
:261.12 g/mol
Boiling Pt:163°C
Specification Test Results
- Appearance:White to Yellow to Orange powder to lump
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together