
4-Butylresorcinol ≥98.0% (by GC)
TCIAB3773-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:50.0 to 55.0 deg-C
:
:166.22 g/mol
:166°C
:GHS07
:18979-61-8
:C₁₀H₁₄O₂
:MFCD01684800