Order Entry
Denmark
ContactUsLinkComponent
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96.0% (by GC)
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96.0% (by GC)
Supplier:  TCI
Certificates
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to lump
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    82.0 to 87.0 deg-C
  • Specific rotation [a]20/D:
    -44.0 to -47.0 deg (C=1, CHCl3)

Specification Test Results

Related Information

Frequently Bought Together