Biotin pentafluorophenyl ester (PFP-Biotin) ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:120550-35-8
:
Formula:C₁₆H₁₅F₅N₂O₃S
MDL Number:MFCD09952636
:410.36 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):6.60 to 7.60 %
- Melting point:190.0 to 194.0 deg-C
- Specific rotation [a]20/D:+39.0 to +43.0 deg (C=1, DMF)
Specification Test Results
Frequently Bought Together