Order Entry
Denmark
ContactUsLinkComponent
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0% (by HPLC)
Health
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0% (by HPLC)
Supplier:  TCI
Certificates
Health
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine ≥99.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +56.0 to +60.0 deg (C=1, MeOH)

Specification Test Results

Related Information

Frequently Bought Together