Order Entry
Denmark
ContactUsLinkComponent
 
2-(4-Bromophenyl)benzimidazole ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
2-(4-Bromophenyl)benzimidazole ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

Frequently Bought Together