2,2'-Biimidazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 2,2'-Diimidazole, 2,2'-Bisimidazole, 1H,1'H-2,2'-Biimidazole
CAS Number:492-98-8
:
Formula:C₆H₆N₄
MDL Number:MFCD00047014
Molecular Weight:134.14 g/mol
Melting Pt:300°C
EINECS:207-768-8
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together