Dimethyl biphenyl-4,4'-dicarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:792-74-5
:
Formula:C₁₆H₁₄O₄
MDL Number:MFCD00017201
:270.28 g/mol
EINECS:212-341-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:217.0 to 220.0 deg-C
Specification Test Results
Frequently Bought Together