
Silver behenate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2489-05-6
:
Formula:C₂₂H₄₄O₂
MDL Number:MFCD00059001
:447.45 g/mol
EINECS:219-641-4
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together