Betanin (red beet extract diluted with dextrin)
Supplier: TCI
Certificates
About this item
CAS Number:7659-95-2
:
Formula:C₂₄H₂₆N₂O₁₃
MDL Number:MFCD00060076
:550.47 g/mol
EINECS:231-628-5
Specification Test Results
- Appearance:Red to Dark red to Dark purple powder to crystal
- Lambda max.:530.0 to 536.0 nm (pH 5.4 buffer sol.)
- Absorbance (E1%1cm):min. 1.6 (pH 5.4 buffer sol., 530.0 to 536.0 nm)
Specification Test Results
Frequently Bought Together