Order Entry
Denmark
ContactUsLinkComponent
 
Benzaldehyde phenylhydrazone ≥98.0% (by HPLC)
  TCIAB0014-25G
 :  TCI
 :  
Benzaldehyde phenylhydrazone ≥98.0% (by HPLC)
  TCIAB0014-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Solubility in hot Methanol:
    almost transparency

 

 

 :
 :196.25 g/mol
 :209-625-5
 :588-64-7
 :C₁₃H₁₂N₂
 :MFCD00051318