Ethyl violet ≥98.0% (by titrimetric analysis) sensitive spectrophotometric reagent for anionic surfactants
TCIAA5101-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Green to Dark green powder to crystal
- Purity (Argentmetric Titration):min. 98.0 % (after drying)
- Purity (Chelometric Titration):min. 98.0 % (after drying)
- Lambda max.:594.0 to 598.0 nm (H2O)
- Absorbance (E1%1cm):min. 1,900 (H2O, 594.0 to 598.0 nm)
:
:492.13 g/mol
:219-231-5
:2390-59-2
:C₃₁H₄₂ClN₃
:MFCD00011765