(R)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3775-73-3
:
Formula:C₄H₉NO₂
MDL Number:MFCD00211284
:103.1198 g/mol
Melting Pt:212°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together