Order Entry
Denmark
ContactUsLinkComponent
Biotin-C5-Amine ≥95.0% (by HPLC)
Biotin-C5-Amine ≥95.0% (by HPLC)
Supplier:  TCI
Certificates
Biotin-C5-Amine ≥95.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 95.0 area%
  • Melting point:
    122.0 to 126.0 deg-C
  • Specific rotation [a]20/D:
    +40.0 to +44.0 deg (C=0.03, MeOH)

Specification Test Results

Frequently Bought Together